α,α,α-Trifluoro-o-toluidine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-(Trifluoromethyl)aniline, 2-Aminobenzotrifluoride
A0433-25ML
A0433-500ML
TCIAA0433-25MLEA
16
EUR
TCIAA0433-25ML
TCIAA0433-500ML
α,α,α-Trifluoro-o-toluidine ≥98.0% (by GC, titration analysis)
α,α,α-Trifluoro-o-toluidine
Formula:
C₇H₆F₃N Peso molecolare: 161,13 g/mol Punto di ebollizione: 171°C Punto di fusione: –33 °C Densità: 1,282 g/cm³ (20 °C) Flash Pt: 55°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007718 CAS Number: 88-17-5 EINECS: 201-806-7 UN: 2942 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid to slightly cloudy liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...