Stampa…

2,3,4,5-Tetrafluorobenzyl alcohol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

T1883-1G T1883-5G
TCIAT1883-1GEA 28 EUR
TCIAT1883-1G TCIAT1883-5G
2,3,4,5-Tetrafluorobenzyl alcohol ≥98.0% (by GC)
2,3,4,5-Tetrafluorobenzyl alcohol
Formula: C₇H₄F₄O
Peso molecolare: 180,1 g/mol
Flash Pt: 101°C
MDL Number: MFCD01631452
CAS Number: 53072-18-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=966e508266c1d651a44cf71ec83c9bdfb9caa417&vid=966e508266c1d651a44cf71ec83c9bdfb9caa417&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR