Stampa…

Didodecylamine ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Dilaurylamine

D0267-25G D0267-5G
TCIAD0267-25GEA 246 EUR
TCIAD0267-25G TCIAD0267-5G
Didodecylamine ≥97.0% (by titrimetric analysis)
Didodecylamine
Formula: C₂₄H₅₁N
Peso molecolare: 353,68 g/mol
Punto di ebollizione: 265°C
Punto di fusione: 100…102 °C
MDL Number: MFCD00041918
CAS Number: 3007-31-6
EINECS: 221-114-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91582d37506aecc475ef5a56120607fd9f612a33&vid=91582d37506aecc475ef5a56120607fd9f612a33&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 48.0 to 55.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR