α-Bromo-4-(trifluoromethoxy)toluene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
1-(Bromomethyl)-4-(trifluoromethoxy)benzene, p-(Trifluoromethoxy)benzyl bromide, 4-(Trifluoromethoxy)benzyl bromide
T1781-1G
T1781-5G
TCIAT1781-1GEA
24
EUR
TCIAT1781-1G
TCIAT1781-5G
α-Bromo-4-(trifluoromethoxy)toluene ≥98.0% (by GC)
α-Bromo-4-(trifluoromethoxy)toluene
Formula:
C₈H₆BrF₃O Peso molecolare: 255,03 g/mol Punto di ebollizione: 79°C Punto di fusione: 23…24 °C Densità: 1,58 g/cm³ (20 °C) |
MDL Number:
MFCD00061238 CAS Number: 50824-05-0 UN: 3261 ADR: 8,III |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...