Stampa…

2,4,5-Trifluorobenzyl bromide ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1-(Bromomethyl)-2,4,5-trifluorobenzene, α-Bromo-2,4,5-trifluorotoluene

T2386-1G T2386-5G
TCIAT2386-1GEA 30 EUR
TCIAT2386-1G TCIAT2386-5G
2,4,5-Trifluorobenzyl bromide ≥98.0% (by GC, titration analysis)
2,4,5-Trifluorobenzyl bromide
Formula: C₇H₄BrF₃
Peso molecolare: 225,01 g/mol
Punto di ebollizione: 182 °C (1013 hPa)
Punto di fusione: 21 °C
MDL Number: MFCD00061209
CAS Number: 157911-56-3
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=955c2b8d1a3fafc7a43a9724214938771fa94319&vid=955c2b8d1a3fafc7a43a9724214938771fa94319&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR