3,5-Difluorobenzylamine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
D3343-1G
D3343-5G
TCIAD3343-1GEA
27
EUR
TCIAD3343-1G
TCIAD3343-5G
3,5-Difluorobenzylamine ≥98.0% (by GC, titration analysis)
3,5-Difluorobenzylamine
Formula:
C₇H₇F₂N Peso molecolare: 143,14 g/mol Punto di ebollizione: 54 °C (1 -rr) Densità: 1,21 g/cm³ (20 °C) Flash Pt: 74°C |
MDL Number:
MFCD00061244 CAS Number: 90390-27-5 UN: 2735 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...