2-Fluorobenzylamine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
F0538-25G
F0538-5G
TCIAF0538-25GEA
117
EUR
TCIAF0538-25G
TCIAF0538-5G
2-Fluorobenzylamine ≥98.0% (by GC, titration analysis)
2-Fluorobenzylamine
Formula:
C₇H₈FN Peso molecolare: 125,15 g/mol Punto di ebollizione: 75°C Densità: 1,09 g/cm³ (20 °C) Flash Pt: 67°C |
MDL Number:
MFCD00008107 CAS Number: 89-99-6 EINECS: 201-957-9 UN: 2735 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...