Suberic acid ≥99.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1,8-Octanedioic acid, Octanedioic acid, 1,6-Hexanedicarboxylic acid
O0023-25G
O0023-500G
O0023-100G
TCIAO0023-25GEA
16
EUR
TCIAO0023-25G
TCIAO0023-500G
TCIAO0023-100G
Suberic acid ≥99.0% (by GC, titration analysis)
Suberic acid
Formula:
C₈H₁₄O₄ Peso molecolare: 174,2 g/mol Punto di ebollizione: 279°C Punto di fusione: 139…142 °C Densità: 1,354 g/cm³ (21 °C) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00004428 CAS Number: 505-48-6 EINECS: 208-010-9 Merck Index: 14,08862 |
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 141.0 to 144.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...