Dimethyl glutarate ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
1,5-Pentanedioic acid dimethyl ester, Glutaric acid dimethyl ester
G0185-25G
G0185-500G
G0185-100G
TCIAG0185-25GEA
16
EUR
TCIAG0185-25G
TCIAG0185-500G
TCIAG0185-100G
Dimethyl glutarate ≥98.0% (by GC)
Dimethyl glutarate
Formula:
C₇H₁₂O₄ Peso molecolare: 160.17 g/mol Punto di ebollizione: 214°C Punto di fusione: -37 Flash Pt: 110°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008468 CAS Number: 1119-40-0 EINECS: 214-277-2 Merck Index: 13,04486 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...