Stampa…

Tetrafluoroterephthalonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Perfluoroterephthalonitrile

T1050-1G T1050-25G T1050-5G
TCIAT1050-1GEA 16 EUR
TCIAT1050-1G TCIAT1050-25G TCIAT1050-5G
Tetrafluoroterephthalonitrile ≥98.0% (by GC)
Tetrafluoroterephthalonitrile
Formula: C₈F₄N₂
Peso molecolare: 200,1 g/mol
MDL Number: MFCD00001776
CAS Number: 1835-49-0
EINECS: 217-397-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e7b0d33ae7235bd3c7dd55f56289adcebdc4c77&vid=2e7b0d33ae7235bd3c7dd55f56289adcebdc4c77&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 196.0 to 200.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR