2,4-Dichlorophenyl isocyanate ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2,4-Dichlorophenylisocyantae
I0555-25G
I0555-5G
TCIAI0555-25GEA
136
EUR
TCIAI0555-25G
TCIAI0555-5G
2,4-Dichlorophenyl isocyanate ≥98.0% (by GC, titration analysis)
2,4-Dichlorophenyl isocyanate
Formula:
C₇H₃Cl₂NO Peso molecolare: 188,01 g/mol Punto di ebollizione: 235 °C (1013 hPa) Punto di fusione: 59…61 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00002001 CAS Number: 2612-57-9 EINECS: 220-040-4 UN: 2250 ADR: 6.1,II |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Butylamine Method) | min. 98.0 % |
Melting point | 57.0 to 60.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...