L(+)-Leucina ≥99.0% (by titrimetric analysis)
Fornitore: TCI
Synonyms:
H-Leu-OH, (S)-(+)-Leucina, Leu, L-Leucina
Formula:
C₆H₁₃NO₂ Peso molecolare: 131.17 g/mol Punto di fusione: 300 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00002617 CAS Number: 61-90-5 EINECS: 200-522-0 Merck Index: 14,05451 |
|
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Specific rotation [a]20/D | +14.7 to +15.7 deg (C=4, HCl (1+1)) |
Solubility in dilute HCl | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...