D-(-)-Leucine ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
H-D-Leu-OH, (R)-(-)-Leucine
L0027-25G
L0027-5G
TCIAL0027-25GEA
69
EUR
TCIAL0027-25G
TCIAL0027-5G
D-(-)-Leucine ≥98.0% (by titrimetric analysis)
D-(-)-Leucine
Formula:
C₆H₁₃NO₂ Peso molecolare: 131,17 g/mol Punto di ebollizione: 226 °C (1013 hPa) |
MDL Number:
MFCD00063088 CAS Number: 328-38-1 EINECS: 206-327-7 |
Risultati del test di specificazione
Appearance | White powder to crystaline |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -13.5 to -15.5 deg (C=4, 6mol/L HCl) |
Solubility in dilute HCl | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...