(Methyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
M0779-100G
M0779-25G
M0779-500G
TCIAM0779-100GEA
55
EUR
TCIAM0779-100G
TCIAM0779-25G
TCIAM0779-500G
(Methyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
(Methyl)triphenylphosphonium bromide
Formula:
C₁₉H₁₈BrP Peso molecolare: 357,23 g/mol Punto di fusione: 232 °C Flash Pt: ≥ 240 °C |
MDL Number:
MFCD00011804 CAS Number: 1779-49-3 EINECS: 217-218-9 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 233.0 to 238.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...