(4-Methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
4-Methoxyphenylacetic acid
Formula:
(C₆H₄(OCH₃))CH₂COOH Peso molecolare: 166.18 g/mol Punto di ebollizione: 305°C Flash Pt: 193 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00004345 CAS Number: 104-01-8 EINECS: 203-166-4 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 85.0 to 88.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...