Stampa…

(2,5-Dimethoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

D1445-25G D1445-5G
TCIAD1445-25GEA 145 EUR
TCIAD1445-25G TCIAD1445-5G
(2,5-Dimethoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2,5-Dimethoxyphenyl)acetic acid
Formula: (C₆H₃(OCH₃)₂)CH₂COOH
Peso molecolare: 196,2 g/mol
Punto di ebollizione: 327 °C (1013 hPa)
Punto di fusione: 123 °C
MDL Number: MFCD00004322
CAS Number: 1758-25-4
EINECS: 217-151-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9626f3245c4e6aac999b2cee0ab8f6560cc0800&vid=c9626f3245c4e6aac999b2cee0ab8f6560cc0800&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 123.0 to 126.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR