9-Fluorenylmethanol ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₄H₁₂O Punto di ebollizione: 337 °C (1013 hPa) Punto di fusione: 107 °C |
MDL Number:
MFCD00001139 CAS Number: 24324-17-2 EINECS: 246-167-5 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 103.0 to 106.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...