Stampa…

(S)-(+)-2-Methylpiperazine ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (S)-2-Methylpiperazine

M1424-1G M1424-5G
TCIAM1424-1GEA 34 EUR
TCIAM1424-1G TCIAM1424-5G
(S)-(+)-2-Methylpiperazine ≥97.0% (by titrimetric analysis)
(S)-(+)-2-Methylpiperazine
Formula: C₅H₁₂N₂
Peso molecolare: 100,16 g/mol
Punto di ebollizione: 155 - 156 °C (1013 hPa)
MDL Number: MFCD00171452
CAS Number: 74879-18-8
UN: 2925
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42307b31dc612fe79ae889dfac6dc748034a43d0&vid=42307b31dc612fe79ae889dfac6dc748034a43d0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Optical purity (GC) min. 99.0 ee%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 88.0 to 92.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR