Stampa…

4-Cyclohexylcyclohexanol cis- and trans-mixture ≥98.0% (by GC)

Fornitore: TCI EUROPE

C0497-25G C0497-5G
TCIAC0497-25GEA 129 EUR
TCIAC0497-25G TCIAC0497-5G
4-Cyclohexylcyclohexanol cis- and trans-mixture ≥98.0% (by GC)
4-Cyclohexylcyclohexanol
Formula: C₁₂H₂₂O
Peso molecolare: 182,31 g/mol
MDL Number: MFCD00060071
CAS Number: 2433-14-9
EINECS: 219-418-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8908469ea600aca405b87099e92df104f872e5ff&vid=439fbbd89753ad5dcda5deb3e4aa07509eae96bb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 % (total of isomer)
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR