Dimethyl sulphite ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl sulphite, Dimethylsulphite
D0264-25G
D0264-500G
TCIAD0264-25GEA
81
EUR
TCIAD0264-25G
TCIAD0264-500G
Dimethyl sulphite ≥99.0% (by GC)
Dimethyl sulphite
Formula:
C₂H₆O₃S Peso molecolare: 110,13 g/mol Punto di ebollizione: 126°C Densità: 1,29 g/cm³ (20 °C) Flash Pt: 30°C |
MDL Number:
MFCD00008415 CAS Number: 616-42-2 EINECS: 210-481-0 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...