5-Nitroindole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Nitro-1H-indole
N0400-25G
N0400-5G
TCIAN0400-25GEA
206
EUR
TCIAN0400-25G
TCIAN0400-5G
5-Nitroindole ≥98.0% (by GC)
5-Nitroindole
Formula:
C₈H₆N₂O₂ Peso molecolare: 162,15 g/mol Punto di fusione: 140 °C |
MDL Number:
MFCD00005673 CAS Number: 6146-52-7 EINECS: 228-153-0 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 140.0 to 144.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...