N-Ethylethylenediamine ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Aminoethyl(ethyl)amine
E0294-25ML
E0294-500ML
E0294-100ML
TCIAE0294-25MLEA
27
EUR
TCIAE0294-25ML
TCIAE0294-500ML
TCIAE0294-100ML
N-Ethylethylenediamine ≥99.0% (by GC)
N-Ethylethylenediamine
Formula:
C₄H₁₂N₂ Peso molecolare: 88,15 g/mol Punto di ebollizione: 130°C Densità: 0,83 g/cm³ (20 °C) Flash Pt: 33°C |
MDL Number:
MFCD00008166 CAS Number: 110-72-5 EINECS: 203-795-4 UN: 2734 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...