2-Methoxybenzylamine ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(2-Methoxyphenyl)methanamine, 2-(Aminomethyl)anisole, o-Anisylamine
M0869-25ML
M0869-5ML
TCIAM0869-25MLEA
65
EUR
TCIAM0869-25ML
TCIAM0869-5ML
2-Methoxybenzylamine ≥97.0% (by GC, titration analysis)
2-Methoxybenzylamine
Formula:
C₈H₁₁NO Peso molecolare: 137,18 g/mol Punto di ebollizione: 127°C Densità: 1,05 g/cm³ (20 °C) Flash Pt: 100°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00008110 CAS Number: 6850-57-3 EINECS: 229-941-7 UN: 2735 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...