Stampa…

2-Amino-4-nitrophenol ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Hydroxy-5-nitroaniline

A0378-25G A0378-500G A0378-100G
TCIAA0378-25GEA 64 EUR
TCIAA0378-25G TCIAA0378-500G TCIAA0378-100G
2-Amino-4-nitrophenol ≥98.0% (by titrimetric analysis)
2-Amino-4-nitrophenol
Formula: C₆H₆N₂O₃
Peso molecolare: 154,13 g/mol
Punto di fusione: 176…177 °C
Densità: 1,66 g/cm³ (20 °C)
MDL Number: MFCD00007695
CAS Number: 99-57-0
EINECS: 202-767-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e25f230a918ca117e53bc7aa2c05d8b27f178a55&vid=e25f230a918ca117e53bc7aa2c05d8b27f178a55&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 144.0 to 148.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR