2-Amino-5-nitropyridine ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Nitropyridin-2-ylamine, 5-Nitropyridin-2-amine, 5-Nitro-2-pyridylamine
A0794-25G
A0794-250G
A0794-5G
TCIAA0794-25GEA
41
EUR
TCIAA0794-25G
TCIAA0794-250G
TCIAA0794-5G
2-Amino-5-nitropyridine ≥98.0% (by titrimetric analysis)
2-Amino-5-nitropyridine
Formula:
C₅H₅N₃O₂ Peso molecolare: 139,11 g/mol Punto di fusione: 185…188 °C Densità: 1,623 g/cm³ (20 °C) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00006325 CAS Number: 4214-76-0 EINECS: 224-145-6 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 185.0 to 190.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...