2-Amino-4-methylpyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Aminopicoline, 2-Amino-4-picoline, 4-Methyl-2-pyridylamine, 4-Methylpyridin-2-amine
A0402-25G
A0402-500G
TCIAA0402-25GEA
34
EUR
TCIAA0402-25G
TCIAA0402-500G
2-Amino-4-methylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-4-methylpyridine
Formula:
C₆H₈N₂ Peso molecolare: 108,14 g/mol Punto di ebollizione: 230,9 °C (1013 hPa) Punto di fusione: 98 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00006321 CAS Number: 695-34-1 EINECS: 211-780-9 UN: 2811 ADR: 6.1,III Merck Index: 14,00466 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 98.0 to 101.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...