5-Aminoindole ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Indol-5-ylamine, 5-Indolamine, 5-Amino-1H-indole, 1H-Indol-5-amine
A2071-1G
A2071-5G
TCIAA2071-1GEA
35
EUR
TCIAA2071-1G
TCIAA2071-5G
5-Aminoindole ≥98.0% (by GC, titration analysis)
5-Aminoindole
Formula:
C₈H₈N₂ Peso molecolare: 132,16 g/mol Punto di ebollizione: 190°C Punto di fusione: 131…134 °C |
MDL Number:
MFCD00005679 CAS Number: 5192-03-0 EINECS: 225-977-2 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...