Stampa…

3-Methylbenzylamine ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: m-Tolylmethanamine, 3-(Aminomethyl)toluene

M1274-1ML M1274-5ML
TCIAM1274-1MLEA 20 EUR
TCIAM1274-1ML TCIAM1274-5ML
3-Methylbenzylamine ≥97.0% (by GC, titration analysis)
3-Methylbenzylamine
Formula: C₈H₁₁N
Peso molecolare: 121,18 g/mol
Punto di ebollizione: 206°C
Densità: 0,9809 g/cm³ (0 °C)
Flash Pt: 81°C
MDL Number: MFCD00008118
CAS Number: 100-81-2
EINECS: 202-890-8
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6af4bc70046eff5c1501f8641e14566c5d8eed43&vid=6af4bc70046eff5c1501f8641e14566c5d8eed43&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR