1-Iodooctadecane ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Stearyl Iodide, Octadecyl Iodide
I0065-25G
I0065-100G
TCIAI0065-25GEA
98
EUR
TCIAI0065-25G
TCIAI0065-100G
1-Iodooctadecane ≥98.0% (by GC)
1-Iodooctadecane
Formula:
C₁₈H₃₇I Peso molecolare: 380,4 g/mol Punto di ebollizione: 197°C Punto di fusione: 34 °C Densità: 1,104 g/cm³ (20 °C) |
MDL Number:
MFCD00001090 CAS Number: 629-93-6 EINECS: 211-117-3 |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Solubility in Toluene | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...