Stampa…

3-(Trifluoromethyl)thiophenol ≥95.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: 3-Mercaptobenzotrifluoride, 3-(Trifluoromethyl)benzenethiol, 3-(Trifluoromethyl)benzene-1-thiol

T2780-25G T2780-5G
TCIAT2780-25GEA 352 EUR
TCIAT2780-25G TCIAT2780-5G
3-(Trifluoromethyl)thiophenol ≥95.0% (by GC, titration analysis)
3-(Trifluoromethyl)thiophenol
Formula: C₇H₅F₃S
Punto di ebollizione: 163°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00041142
CAS Number: 937-00-8
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc1e57a9c6a94087cef20cf7d4152f639929e55c&vid=bc1e57a9c6a94087cef20cf7d4152f639929e55c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR