Citraconic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Methyl-2-butenedioic acid
C0363-25G
C0363-500G
TCIAC0363-25GEA
20
EUR
TCIAC0363-25G
TCIAC0363-500G
Citraconic acid ≥98.0% (by GC, titration analysis)
Citraconic acid
Formula:
C₅H₆O₄ Peso molecolare: 130,1 g/mol Punto di ebollizione: 336 °C (1013 hPa) Punto di fusione: 98…99 °C Densità: 1,617 g/cm³ (20 °C) |
MDL Number:
MFCD00078086 CAS Number: 498-23-7 EINECS: 207-858-7 Merck Index: 14,02321 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Water | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...