3-Methoxycinnamic acid ≥99.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(3-Methoxyphenyl)acrylic acid
M0444-25G
M0444-250G
TCIAM0444-25GEA
60
EUR
TCIAM0444-25G
TCIAM0444-250G
3-Methoxycinnamic acid ≥99.0% (by GC, titration analysis)
3-Methoxycinnamic acid
Formula:
C₁₀H₁₀O₃ Peso molecolare: 178,19 g/mol |
MDL Number:
MFCD00004386 CAS Number: 6099-04-3 EINECS: 228-049-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 117.0 to 120.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...