Benzofenone ≥99.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Diphenylmethanone, Diphenyl ketone
Formula:
C₁₃H₁₀O Peso molecolare: 182.22 g/mol Punto di ebollizione: 305°C Flash Pt: 150 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00003076 CAS Number: 119-61-9 EINECS: 204-337-6 UN: 3077 ADR: 9 III Merck Index: 14,01098 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 48.0 to 50.0 deg-C |
Solubility in EtOH | almost transparency |
Ignition residue (Sulfate) | max. 0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...