Indole-4-carbonitrile ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
4-Cyano-1H-indole, 1H-Indole-4-carbonitrile, 4-Cyanoindole
C2083-1G
C2083-5G
TCIAC2083-1GEA
25
EUR
TCIAC2083-1G
TCIAC2083-5G
Indole-4-carbonitrile ≥98.0% (by GC)
Indole-4-carbonitrile
Formula:
C₉H₆N₂ Peso molecolare: 142,16 g/mol |
MDL Number:
MFCD00152045 CAS Number: 16136-52-0 UN: 3439 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 118.0 and le 123.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...