Stampa…
2-Chloro-5-iodobenzoic acid ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
C2168-25G C2168-5G
TCIAC2168-25GEA 194 EUR
TCIAC2168-25G TCIAC2168-5G
2-Chloro-5-iodobenzoic acid ≥97.0% (by GC, titration analysis)
2-Chloro-5-iodobenzoic acid
Formula: C₇H₄ClIO₂
Peso molecolare: 282,46 g/mol
MDL Number: MFCD00079731
CAS Number: 19094-56-5
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8109800504f0127b6f34e3201b2796cd1f68b7c&vid=b8109800504f0127b6f34e3201b2796cd1f68b7c&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 158.0 to 162.0 deg-C