4-Chloro-3-nitrocinnamic acid ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(4-Chloro-3-nitrophenyl)prop-2-enoic acid, 3-(4-Chloro-3-nitrophenyl)acrylic acid
C1205-25G
C1205-5G
TCIAC1205-25GEA
273
EUR
TCIAC1205-25G
TCIAC1205-5G
4-Chloro-3-nitrocinnamic acid ≥97.0% (by titrimetric analysis)
4-Chloro-3-nitrocinnamic acid
Formula:
C₉H₆ClNO₄ Peso molecolare: 227,601 g/mol |
MDL Number:
MFCD00063311 CAS Number: 20797-48-2 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 187.0 to 190.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...