Stampa…

6-Chloro-m-toluic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

C1780-1G C1780-5G
TCIAC1780-1GEA 39 EUR
TCIAC1780-1G TCIAC1780-5G
6-Chloro-m-toluic acid ≥98.0% (by GC, titration analysis)
6-Chloro-m-toluic acid
Formula: C₈H₇ClO₂
Peso molecolare: 170,6 g/mol
Punto di fusione: 150°C
MDL Number: MFCD00045798
CAS Number: 6342-60-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d84a1f42f98e12f2537db69391df08c2f15646e7&vid=d84a1f42f98e12f2537db69391df08c2f15646e7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 147.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR