Stampa…

2-Bromo-5-hydroxypyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Bromo-5-pyridinol, 6-Bromopyridin-3-ol

B3657-1G B3657-5G
TCIAB3657-1GEA 22 EUR
TCIAB3657-1G TCIAB3657-5G
2-Bromo-5-hydroxypyridine ≥98.0% (by GC, titration analysis)
2-Bromo-5-hydroxypyridine
Formula: C₅H₄BrNO
Peso molecolare: 174 g/mol
MDL Number: MFCD04114184
CAS Number: 55717-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc62f20f923c374ef7f8d9ab7e9ee9d2dc84e1ca&vid=bc62f20f923c374ef7f8d9ab7e9ee9d2dc84e1ca&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR