2-Bromo-3-fluoropyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B3666-1G
B3666-5G
TCIAB3666-1GEA
49
EUR
TCIAB3666-1G
TCIAB3666-5G
2-Bromo-3-fluoropyridine ≥98.0% (by GC)
2-Bromo-3-fluoropyridine
Formula:
C₅H₃BrFN Punto di ebollizione: 62°C |
MDL Number:
MFCD04114128 CAS Number: 40273-45-8 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...