Stampa…

(±)-2-Bromopropionic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: DL-2-Bromopropanoic acid, 2-Bromopropanoic acid, DL-2-Bromopropionic acid, (±)-2-Bromopropanoic acid

B0644-25G B0644-500G
TCIAB0644-25GEA 18 EUR
TCIAB0644-25G TCIAB0644-500G
(±)-2-Bromopropionic acid ≥98.0% (by GC, titration analysis)
(±)-2-Bromopropionic acid
Formula: H₃CBrCHCOOH
Peso molecolare: 152,98 g/mol
Punto di ebollizione: 205°C
Punto di fusione: ∼25 °C
Densità: 1,7 g/cm³ (25 °C)
Flash Pt: 100°C
MDL Number: MFCD00004211
CAS Number: 598-72-1
EINECS: 209-947-6
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98f18e377f3516e0203fef4ae6957f73b1c986e3&vid=98f18e377f3516e0203fef4ae6957f73b1c986e3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR