Stampa…

5-Bromovaleryl chloride ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

B2059-25G B2059-250G B2059-5G
TCIAB2059-25GEA 110 EUR
TCIAB2059-25G TCIAB2059-250G TCIAB2059-5G
5-Bromovaleryl chloride ≥98.0% (by titrimetric analysis)
5-Bromovaleryl chloride
Formula: BrH₂C(CH₂)₃COOCl
Punto di ebollizione: 118°C
Flash Pt: 107°C
MDL Number: MFCD00013660
CAS Number: 4509-90-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6115efa5a9999afcf19fdc332e420bbe12801eb&vid=d6115efa5a9999afcf19fdc332e420bbe12801eb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR