2-Phenylindole ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
2-Phenyl-1H-indole
P0188-25G
P0188-250G
TCIAP0188-25GEA
16
EUR
TCIAP0188-25G
TCIAP0188-250G
2-Phenylindole ≥98.0% (by GC)
2-Phenylindole
Formula:
C₁₄H₁₁N Peso molecolare: 193.25 g/mol Punto di ebollizione: 420 |
MDL Number:
MFCD00005608 CAS Number: 948-65-2 EINECS: 213-436-3 |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 188.0 to 193.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...