Benzimidazole ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
1H-Benzo[d]imidazole, 1H-Benzimidazole
B0054-25G
B0054-500G
B0054-100G
TCIAB0054-25GEA
16
EUR
TCIAB0054-25G
TCIAB0054-500G
TCIAB0054-100G
Benzimidazole ≥98.0% (by GC, titration analysis)
Benzimidazole
Formula:
C₇H₆N₂ Peso molecolare: 118,14 g/mol Punto di ebollizione: 360 °C (1013 hPa) Punto di fusione: 170…173 °C Densità: 1,23 g/cm³ (20 °C) |
MDL Number:
MFCD00005585 CAS Number: 51-17-2 EINECS: 200-081-4 Merck Index: 14,01081 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 170.0 to 176.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...