Ethyl methanesulphonate ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
M0607-25G
M0607-100G
TCIAM0607-25GEA
37
EUR
TCIAM0607-25G
TCIAM0607-100G
Ethyl methanesulphonate ≥99.0% (by GC)
Ethyl methanesulphonate
Formula:
C₃H₈O₃S Peso molecolare: 124,16 g/mol Punto di ebollizione: 89°C Densità: 1,2079 g/cm³ (20 °C) Flash Pt: 100°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007559 CAS Number: 62-50-0 EINECS: 200-536-7 ADR: 6.1,II Merck Index: 14,03827 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...