Cyclohexyltrimethoxysilane ≥97.0% (by GC)
Fornitore: TCI
|
Attenzione
|
C2280-25G
C2280-5G
TCIAC2280-25GEA
182
EUR
TCIAC2280-25G
TCIAC2280-5G
Cyclohexyltrimethoxysilane ≥97.0% (by GC)
Cyclohexyltrimethoxysilane
Formula:
C₉H₂₀O₃Si Punto di ebollizione: 208°C |
MDL Number:
MFCD01074466 CAS Number: 17865-54-2 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...