Dibutyltin oxide ≥95.0% (W)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide
D0305-25G
D0305-100G
TCIAD0305-25GEA
18
EUR
TCIAD0305-25G
TCIAD0305-100G
Dibutyltin oxide ≥95.0% (W)
Dibutyltin oxide
Formula:
C₈H₁₈OSn Peso molecolare: 248,94 g/mol Punto di fusione: > 300 °C Densità: 1,6 g/cm³ (20 °C) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00001992 CAS Number: 818-08-6 EINECS: 212-449-1 UN: 3146 ADR: 6.1,II |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with ignition residue) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...