Stampa…

Bis(4-nitrophenyl)methane ≥99.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4,4-Dinitrodiphenylmethane, 4,4'-Dinitrodiphenylmethane

D0833-1G D0833-5G
TCIAD0833-1GEA 69 EUR
TCIAD0833-1G TCIAD0833-5G
Bis(4-nitrophenyl)methane ≥99.0% (by GC)
Bis(4-nitrophenyl)methane
Formula: C₁₃H₁₀N₂O₄
Peso molecolare: 258,23 g/mol
MDL Number: MFCD00059184
CAS Number: 1817-74-9
EINECS: 217-330-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3128661f096f3445928f8cb29049ce8342a5e5aa&vid=3128661f096f3445928f8cb29049ce8342a5e5aa&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 187.0 to 191.0 deg-C
Solubility in hot Chloroform almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR