Stampa…

4-Nitroindole ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-Nitro-1H-indole

N0628-1G N0628-5G
TCIAN0628-1GEA 31 EUR
TCIAN0628-1G TCIAN0628-5G
4-Nitroindole ≥98.0% (by GC)
4-Nitroindole
Formula: C₈H₆N₂O₂
Peso molecolare: 162,15 g/mol
Punto di fusione: 206…209 °C
MDL Number: MFCD00010056
CAS Number: 4769-97-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=302cc716c26363b8b8538fb1f441c0bb036a3182&vid=302cc716c26363b8b8538fb1f441c0bb036a3182&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 203.0 to 208.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR