Bario difenilammin-4-solfonato ≥98.0% (by total nitrogen basis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Diphenylamine-4-sulphonic acid barium salt, N-Phenylsulphanilic acid barium salt
Formula:
C₂₄H₂₀BaN₂O₆S₂ Punto di fusione: 300 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007497 CAS Number: 6211-24-1 EINECS: 228-278-0 UN: 1564 ADR: 6.1,III Merck Index: 14,07313 |
|
Risultati del test di specificazione
Appearance | White to Light gray to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Drying loss | max. 1.0 % |
Sensitiveness | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...