2-Amino-5-nitrophenol ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Hydroxy-4-nitroaniline
N0113-25G
N0113-500G
TCIAN0113-25GEA
25
EUR
TCIAN0113-25G
TCIAN0113-500G
2-Amino-5-nitrophenol ≥98.0% (by GC, titration analysis)
2-Amino-5-nitrophenol
Formula:
C₆H₆N₂O₃ Peso molecolare: 154,13 g/mol Punto di fusione: 207…208 °C Densità: 1,54 g/cm³ (20 °C) |
MDL Number:
MFCD00007692 CAS Number: 121-88-0 EINECS: 204-503-8 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 201.0 to 205.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...